Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Ammoniumchlorid

Ammoniumchlorid

25 kg
Katalog # ROTHP726.5
Irriterende
Marketsource
Leverandør:   Roth Carl
Ammoniumchlorid
Katalog # ROTHP726.5
Leverandør:  Roth Carl
Marketsource
CAS nummer:  12125-02-9
Ammoniumchlorid
Ammoniumchlorid

25 kg

CAS nummer:  12125-02-9
Price
2.277,63 kr
Log ind for tilgængelighed
1 * 25 kg
Product Details & Documents
Synonymer:  Ammonium muriate, Salmiac
  • CAS nummer:12125-02-9
  • Formel:NH₄Cl
  • MDL nummer:MFCD00011420
  • Molecular Weight:53.49 g/mol
  • Merck Index:13,00510
  • Smeltende Pt:335 °C
  • Tæthed:1,5256 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:235-186-4
Specifikationer

Specifikationer

  • Pakn:
    25 kg

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.