Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Ammoniumchlorid

Glass bottle for solids, 1 kg
Katalog # ROTHP726.2
Irriterende
Marketsource
Leverandør:   Roth Carl
Ammoniumchlorid
Katalog # ROTHP726.2
Leverandør:  Roth Carl
Marketsource
CAS nummer:  12125-02-9
Ammoniumchlorid
Ammoniumchlorid

Glass bottle for solids, 1 kg

CAS nummer:  12125-02-9
Price
309,91 kr
Log ind for tilgængelighed
1 * 1 kg
Product Details & Documents
Synonymer:  Ammonium muriate, Salmiac
  • CAS nummer:12125-02-9
  • Formel:NH₄Cl
  • MDL nummer:MFCD00011420
  • Molecular Weight:53.49 g/mol
  • Merck Index:13,00510
  • Smeltende Pt:335 °C
  • Tæthed:1,5256 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:235-186-4
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    1 kg

Specifikationer