Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Ammoniumchlorid

Ammoniumchlorid

Glass bottle for solids, 500 g
Katalog # ROTHP726.1
Irriterende
Marketsource
Leverandør:   Roth Carl
Ammoniumchlorid
Katalog # ROTHP726.1
Leverandør:  Roth Carl
Marketsource
CAS nummer:  12125-02-9
Ammoniumchlorid
Ammoniumchlorid

Glass bottle for solids, 500 g

CAS nummer:  12125-02-9
Price
182,96 kr
Log ind for tilgængelighed
1 * 500 g
Product Details & Documents
Synonymer:  Ammonium muriate, Salmiac
  • CAS nummer:12125-02-9
  • Formel:NH₄Cl
  • MDL nummer:MFCD00011420
  • Molecular Weight:53.49 g/mol
  • Merck Index:13,00510
  • Smeltende Pt:335 °C
  • Tæthed:1,5256 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:235-186-4
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    500 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.