Ampicillin, natriumsalt
Glass bottle for solids, 10 g
Katalog # ROTHK029.1
Marketsource
Leverandør: Roth Carl
Gå til:
Product Details & Documents
Synonymer: D(-)-α-Aminobenzylpenicillin Sodium Salt, Ampicillin Na, Ampicillin sodium salt, D-(−)-α-Aminobenzylpenicillin sodium salt Ampicillin sodium salt
- CAS nummer:69-52-3
- Formel:C₁₆H₁₈N₃NaO₄S
- MDL nummer:MFCD00064313
- Molecular Weight:371.39 g/mol
- Opbevaringstemperatur:Køleskab
- EINECS:200-708-1
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:10 g



