Orders LinkContactUsLinkComponent
Diethyl pyrocarbonat
Diethyl pyrocarbonat
Glass bottle, 25 mL
Catalog # ROTHK028.1
Irritant
Marketsource
Supplier:   Roth Carl
Diethyl pyrocarbonat
Catalog # ROTHK028.1
Supplier:  Roth Carl
Marketsource
CAS Number:  1609-47-8
Diethyl pyrocarbonat
Glass bottle, 25 mL
CAS Number:  1609-47-8
Price
596,66 kr
Log in for availability
1 * 25 mL
Product Details & Documents
Synonyms:  DEPC (Diethylpyrocarbonate), Diethyl dicarbonate, DEPC
  • CAS Number:1609-47-8
  • Formula:O(COOC₂H₅)₂
  • MDL Number:MFCD00009106
  • Molecular Weight:162.14 g/mol
  • Merck Index:13,08088
  • Boiling Pt:93 °C (1013 hPa)
  • Density:1,123 g/cm³ (20 °C)
  • Flash Pt:69 °C
  • Storage Temperature:Køleskab
  • EINECS:216-542-8
Specifications
Specifications
  • Pakning:
    Glass bottle
  • Pakn:
    25 mL