Latex gloves, Rotiprotect®
L
Catalog # ROTHE690.1
Marketsource
Supplier: Roth Carl
Latex gloves, Rotiprotect®
L
Jump to:
Product Details & Documents
Sterile, powdered gloves.
Specifications
- Handske størrelse:L
- Seq:03
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
#: 20864.320
Supplier: VWR Chemicals
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
#: 83640.320
Supplier: VWR Chemicals
Acetonitril ≥99.9%, LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
#: 1.00030.2500
Supplier: Merck
Natriumhydroxid 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM Reag. Ph. Eur. volumetric solution
#: 31770.294
Supplier: VWR Chemicals
Vand, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
#: 83645.320
Supplier: VWR Chemicals
Buffer solutions, ready to use, referenced at 20 °C, Certipur®, Supelco®
#: 1.09439.1000
Supplier: Merck
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
#: 20821.365
Supplier: VWR Chemicals
Methanol ≥99.9% (by GC), HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
#: 83638.320
Supplier: VWR Chemicals
Saltsyre 0.1 mol/l (0.1 N), AVS TITRINORM Reag. Ph. Eur. volumetric solution
#: 31955.293
Supplier: VWR Chemicals
Acetonitril ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
#: 20060.320
Supplier: VWR Chemicals



