Orders LinkContactUsLinkComponent
Natriumiodid
Glass bottle for solids, 1 kg
  ROTHA134.2
Marketsource
 :   Roth Carl
Natriumiodid
  ROTHA134.2
 :  Roth Carl
Marketsource
 :  7681-82-5
Natriumiodid
Glass bottle for solids, 1 kg
 :  7681-82-5
6.265,34 kr
1 * 1 kg
 :  Hydriodic acid sodium salt, Sodium monoiodide, Ioduril
  •  :7681-82-5
  •  :NaI
  •  :MFCD00003532
  •  :149.89 g/mol
  •  :13,08705
  •  :1300 °C (1013 hPa)
  •  :662 °C
  •  :3,67 g/cm³ (20 °C)
  •  :Stue temperatur
  •  :231-679-3
  •  :3077
  •  :9,III
  • Pakning:
    Glass bottle for solids
  • Pakn:
    1 kg