Natriumiodid
Glass bottle for solids, 1 kg
ROTHA134.2
Marketsource
: Hydriodic acid sodium salt, Sodium monoiodide, Ioduril
- :7681-82-5
- :NaI
- :MFCD00003532
- :149.89 g/mol
- :13,08705
- :1300 °C (1013 hPa)
- :662 °C
- :3,67 g/cm³ (20 °C)
- :Stue temperatur
- :231-679-3
- :3077
- :9,III
- Pakning:Glass bottle for solids
- Pakn:1 kg