Order Entry
Denmark
Orders LinkContactUsLinkComponent
Natriumiodid

Natriumiodid

Glass bottle for solids, 250 g
Katalog # ROTHA134.1
Miljø
Marketsource
Leverandør:   Roth Carl
Natriumiodid
Katalog # ROTHA134.1
Leverandør:  Roth Carl
Marketsource
CAS nummer:  7681-82-5
Natriumiodid

Glass bottle for solids, 250 g

CAS nummer:  7681-82-5
Price
1.710,55 kr
Log ind for tilgængelighed
1 * 250 g
Product Details & Documents
Synonymer:  Hydriodic acid sodium salt, Sodium monoiodide, Ioduril
  • CAS nummer:7681-82-5
  • Formel:NaI
  • MDL nummer:MFCD00003532
  • Molecular Weight:149.89 g/mol
  • Merck Index:13,08705
  • Kogende Pt:1300 °C (1013 hPa)
  • Smeltende Pt:662 °C
  • Tæthed:3,67 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:231-679-3
  • FN:3077
  • ADR:9,III
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    250 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.