2-Propanol
Glass bottle, 2,5 L
ROTH9866.2
Marketsource
: Isopropylalkohol
- :67-63-0
- :(CH₃)₂CHOH
- :MFCD00011674
- :60.1 g/mol
- :13,05228
- :82 °C (1013 hPa)
- :–89 °C
- :0,786 g/cm³ (20 °C)
- :12 °C
- :Stue temperatur
- :200-661-7
- :1219
- :3,II
- :01-2119457558-25
- Pakning:Glass bottle
- Pakn:2,5 L