Isooctan
Glass bottle, 2,5 L
ROTH9860.3
Marketsource
: Isobutyltrimethylmethane, Isooctane, Isobutyltrimethylethane
- :540-84-1
- :(CH₃)₂CHCH₂C(CH₃)₃
- :MFCD00008943
- :114.23 g/mol
- :13,05212
- :99,2 °C (1013 hPa)
- :–107 °C
- :0,692 g/cm³ (20 °C)
- :–12 °C
- :Stue temperatur
- :208-759-1
- :1262
- :3,II
- Pakning:Glass bottle
- Pakn:2,5 L