Kviksølv
Glass bottle for solids, 100 g
ROTH7594.2
Marketsource
- :7439-97-6
- :Hg
- :MFCD00011035
- :200.59 g/mol
- :13,05925
- :356,5 °C (1013 hPa)
- :–39 °C
- :13,54 g/cm³ (20 °C)
- :Stue temperatur
- :231-106-7
- :2809
- :8,III
- Pakning:Glass bottle for solids
- Pakn:100 g