Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Natriumsilikat

Glass bottle, 1 L
Katalog # ROTH7561.1
Irriterende
Marketsource
Leverandør:   Roth Carl
Natriumsilikat
Katalog # ROTH7561.1
Leverandør:  Roth Carl
Marketsource
CAS nummer:  1344-09-8
Natriumsilikat

Glass bottle, 1 L

CAS nummer:  1344-09-8
Price
133,67 kr
Log ind for tilgængelighed
1 * 1 L
Product Details & Documents
Synonymer:  Vandglas
  • CAS nummer:1344-09-8
  • Formel:Na₂O(SiO₂)x·xH₂O
  • MDL nummer:MFCD00003492
  • Merck Index:13,08751
  • Tæthed:1,39 g/cm³ (25 °C)
  • EINECS:215-687-4
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    1 L

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.