Orders LinkContactUsLinkComponent
Guanin
Guanin
Glass bottle for solids, 10 g
Catalog # ROTH7505.1
Marketsource
Supplier:   Roth Carl
Guanin
Catalog # ROTH7505.1
Supplier:  Roth Carl
Marketsource
CAS Number:  73-40-5
Guanin
Glass bottle for solids, 10 g
CAS Number:  73-40-5
Price
Inquire For Price
Log in for availability
1 * 10 g
Product Details & Documents
Synonyms:  2-Amino-1H-purin-6(7H)-one
  • CAS Number:73-40-5
  • Formula:C₅H₅N₅O
  • MDL Number:MFCD00071533
  • Molecular Weight:151.13 g/mol
  • Merck Index:13,04580
  • Melting Pt:> 220 °C
  • Density:1,67 g/cm³ (20 °C)
  • Storage Temperature:Stue temperatur
  • EINECS:200-799-8
Specifications
Specifications
  • Pakning:
    Glass bottle for solids
  • Pakn:
    10 g