Orders LinkContactUsLinkComponent
Eddikesyre 99% ≥99%
10 L
  ROTH7332.3
Marketsource
 :   Roth Carl
Eddikesyre 99% ≥99%
  ROTH7332.3
 :  Roth Carl
Marketsource
 :  64-19-7
Eddikesyre 99% ≥99%
Eddikesyre 99% ≥99%
10 L
 :  64-19-7
1 * 10 L
 :  Vinegar acid, Methanecarboxylic acid, Ethanoic acid
  •  :64-19-7
  •  :H₃CCOOH
  •  :MFCD00036152
  •  :60.05 g/mol
  •  :Stue temperatur
  •  :200-580-7
  •  :2789
  •  :8,II
  • Pakn:
    10 L