Eddikesyre 99% ≥99%
5 L
Katalog # ROTH7332.2
Marketsource
Leverandør: Roth Carl
Gå til:
Product Details & Documents
Synonymer: Vinegar acid, Methanecarboxylic acid, Ethanoic acid
- CAS nummer:64-19-7
- Formel:H₃CCOOH
- MDL nummer:MFCD00036152
- Molecular Weight:60.05 g/mol
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-580-7
- FN:2789
- ADR:8,II
Specifikationer
Specifikationer
- Pakn:5 L
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



