Eddikesyre 99% ≥99%
Glass bottle, 1 L
Katalog # ROTH7332.1
Marketsource
Leverandør: Roth Carl
Gå til:
Product Details & Documents
Synonymer: Vinegar acid, Methanecarboxylic acid, Ethanoic acid
- CAS nummer:64-19-7
- Formel:H₃CCOOH
- MDL nummer:MFCD00036152
- Molecular Weight:60.05 g/mol
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-580-7
- FN:2789
- ADR:8,II
Specifikationer
Specifikationer
- Pakning:Glass bottle
- Pakn:1 L