Benzen
Glass bottle, 1 L
ROTH7173.1
Marketsource
: Cyclohexatriene
- :71-43-2
- :C₆H₆
- :MFCD00003009
- :78.11 g/mol
- :13,01066
- :80,1 °C (1013 hPa)
- :5,5 °C
- :0,878 g/cm³ (20 °C)
- :–11 °C
- :Stue temperatur
- :200-753-7
- :1114
- :3,II
- :01-2119447106-44
- Pakning:Glass bottle
- Pakn:1 L