Toluen
Plastic bottle, 2,5 L
Katalog # ROTH7115.2
Marketsource
Leverandør: Roth Carl
Gå til:
Product Details & Documents
Synonymer: Methylbenzene, Phenylmethane, FM approval
- CAS nummer:108-88-3
- Formel:C₆H₅CH₃
- MDL nummer:MFCD00008512
- Molecular Weight:92.14 g/mol
- Merck Index:13,09607
- Kogende Pt:110,6 °C (1013 hPa)
- Smeltende Pt:–95 °C
- Tæthed:0,867 g/cm³ (20 °C)
- Flash Pt:4 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:203-625-9
- FN:1294
- ADR:3,II
- NÅ:05-2114615130-69
Specifikationer
Specifikationer
- Pakning:Plastic bottle
- Pakn:2,5 L



