Isooctan
Glass bottle, 2,5 L
Katalog # ROTH6889.2
Marketsource
Leverandør: Roth Carl
Gå til:
Product Details & Documents
Synonymer: Isobutyltrimethylmethane, Isooctane, Isobutyltrimethylethane
- CAS nummer:540-84-1
- Formel:(CH₃)₂CHCH₂C(CH₃)₃
- MDL nummer:MFCD00008943
- Molecular Weight:114.23 g/mol
- Merck Index:13,05212
- Kogende Pt:99,2 °C (1013 hPa)
- Smeltende Pt:–107 °C
- Tæthed:0,692 g/cm³ (20 °C)
- Flash Pt:–12 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:208-759-1
- FN:1262
- ADR:3,II
Specifikationer
Specifikationer
- Pakning:Glass bottle
- Pakn:2,5 L
- Cat. No.:ROTH6889.2



