Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
D(+)-Glucose monohydrat

D(+)-Glucose monohydrat

25 kg
Katalog # ROTH6887.5
Marketsource
Leverandør:   Roth Carl
D(+)-Glucose monohydrat
Katalog # ROTH6887.5
Leverandør:  Roth Carl
Marketsource
CAS nummer:  14431-43-7
D(+)-Glucose monohydrat
D(+)-Glucose monohydrat

25 kg

CAS nummer:  14431-43-7
Price
Spørg for pris
Log ind for tilgængelighed
1 * 25 kg
Product Details & Documents
Synonymer:  (2S,3R,4S,5S,6R)-6-(Hydroxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetraol hydrate, Dextrose monohydrate
  • CAS nummer:14431-43-7
  • Formel:C₆H₁₂O₆·H₂O
  • MDL nummer:MFCD00149450
  • Molecular Weight:198.17 g/mol
  • Smeltende Pt:83 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-075-1
Specifikationer

Specifikationer

  • Pakn:
    25 kg

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.