Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
D(+)-Glucose monohydrat

D(+)-Glucose monohydrat

Glass bottle for solids, 2,5 kg
Katalog # ROTH6887.2
Marketsource
Leverandør:   Roth Carl
D(+)-Glucose monohydrat
Katalog # ROTH6887.2
Leverandør:  Roth Carl
Marketsource
CAS nummer:  14431-43-7
D(+)-Glucose monohydrat
D(+)-Glucose monohydrat

Glass bottle for solids, 2,5 kg

CAS nummer:  14431-43-7
Price
421,92 kr
Log ind for tilgængelighed
1 * 2,5 kg
Product Details & Documents
Synonymer:  (2S,3R,4S,5S,6R)-6-(Hydroxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetraol hydrate, Dextrose monohydrate
  • CAS nummer:14431-43-7
  • Formel:C₆H₁₂O₆·H₂O
  • MDL nummer:MFCD00149450
  • Molecular Weight:198.17 g/mol
  • Smeltende Pt:83 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-075-1
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    2,5 kg

Specifikationer