Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
D(+)-Glucose monohydrat

D(+)-Glucose monohydrat

Glass bottle for solids, 1 kg
Catalog # ROTH6887.1
Marketsource
Supplier:   Roth Carl
D(+)-Glucose monohydrat
Catalog # ROTH6887.1
Supplier:  Roth Carl
Marketsource
CAS Number:  14431-43-7
D(+)-Glucose monohydrat
D(+)-Glucose monohydrat

Glass bottle for solids, 1 kg

CAS Number:  14431-43-7
Price
Inquire For Price
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  (2S,3R,4S,5S,6R)-6-(Hydroxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetraol hydrate, Dextrose monohydrate
  • CAS Number:14431-43-7
  • Formula:C₆H₁₂O₆·H₂O
  • MDL Number:MFCD00149450
  • Molecular Weight:198.17 g/mol
  • Melting Pt:83 °C
  • Storage Temperature:Stue temperatur
  • EINECS:200-075-1
Specifications

Specifications

  • Pakning:
    Glass bottle for solids
  • Pakn:
    1 kg

Specifications