Natriumacetat
Glass bottle for solids, 2,5 kg
ROTH6773.3
Marketsource
: Acetic acid sodium salt
- :127-09-3
- :H₃CCOONa
- :MFCD00012459
- :82.03 g/mol
- :12,08711
- :881,4 °C (1013 hPa)
- :324 °C
- :1,528 g/cm³ (20 °C)
- :>250 °C (closed cup)
- :Stue temperatur
- :204-823-8
- :01-2119485123-42
- Pakning:Glass bottle for solids
- Pakn:2,5 kg