Orders LinkContactUsLinkComponent
Natriumacetat
Glass bottle for solids, 2,5 kg
  ROTH6773.3
Marketsource
 :   Roth Carl
Natriumacetat
  ROTH6773.3
 :  Roth Carl
Marketsource
 :  127-09-3
Natriumacetat
Glass bottle for solids, 2,5 kg
 :  127-09-3
1 * 2,5 kg
 :  Acetic acid sodium salt
  •  :127-09-3
  •  :H₃CCOONa
  •  :MFCD00012459
  •  :82.03 g/mol
  •  :12,08711
  •  :881,4 °C (1013 hPa)
  •  :324 °C
  •  :1,528 g/cm³ (20 °C)
  •  :>250 °C (closed cup)
  •  :Stue temperatur
  •  :204-823-8
  •  :01-2119485123-42
  • Pakning:
    Glass bottle for solids
  • Pakn:
    2,5 kg