Orders LinkContactUsLinkComponent
Citronsyre
Glass bottle for solids, 500 g
  ROTH6490.3
Marketsource
 :   Roth Carl
Citronsyre
  ROTH6490.3
 :  Roth Carl
Marketsource
 :  77-92-9
Citronsyre
Citronsyre
Glass bottle for solids, 500 g
 :  77-92-9
1 * 500 g
 :  2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid
  •  :77-92-9
  •  :HOC(COOH)(CH₂COOH)₂
  •  :MFCD00011669
  •  :192.13 g/mol
  •  :13,02350
  •  :310 °C (1013 hPa)
  •  :149…151 °C
  •  :1,665 g/cm³ (20 °C)
  •  :Stue temperatur
  •  :201-069-1
  •  :01-2119457026-42-0000
  • Pakning:
    Glass bottle for solids
  • Pakn:
    500 g