Citronsyre
Glass bottle for solids, 1 kg
Katalog # ROTH6490.1
Marketsource
Leverandør: Roth Carl
Gå til:
Product Details & Documents
Synonymer: 2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid
- CAS nummer:77-92-9
- Formel:HOC(COOH)(CH₂COOH)₂
- MDL nummer:MFCD00011669
- Molecular Weight:192.13 g/mol
- Merck Index:13,02350
- Kogende Pt:310 °C (1013 hPa)
- Smeltende Pt:149…151 °C
- Tæthed:1,665 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:201-069-1
- NÅ:01-2119457026-42-0000
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:1 kg



