Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Saltsyre 25%

Glass bottle, 1 L
  ROTH6331.1
 
 
Marketsource
 :   Roth Carl
Saltsyre 25%
  ROTH6331.1
 :  Roth Carl
Marketsource
 :  7647-01-0
Saltsyre 25%

Glass bottle, 1 L

 :  7647-01-0
 
 
   
1 * 1 L
 
 :  Hydrochloric acid solution, Muriatic acid
  •  :7647-01-0
  •  :HCl
  •  :MFCD00011324
  •  :36.46 g/mol
  •  :110 °C (1013 hPa)
  •  :–30 °C
  •  :1,18 g/cm³ (20 °C)
  •  :Stue temperatur
  •  :231-595-7
  •  :1789
  •  :8,II
 

 

  • Pakning:
    Glass bottle
  • Pakn:
    1 L