Dichlormethan
Glass bottle, 1 L
Katalog # ROTH6053.1
Marketsource
Leverandør: Roth Carl
Gå til:
Product Details & Documents
Synonymer: Methylenchlorid
- CAS nummer:75-09-2
- Formel:CH₂Cl₂
- MDL nummer:MFCD00000881
- Molecular Weight:84.93 g/mol
- Merck Index:12,06140
- Kogende Pt:39,8 °C (1013 hPa)
- Smeltende Pt:–95 °C
- Tæthed:1,322 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-838-9
- FN:1593
- ADR:6.1,III
Specifikationer
Specifikationer
- Pakning:Glass bottle
- Pakn:1 L