Ponceau S
Glass bottle for solids, 10 g
Katalog # ROTH5938.2
Marketsource
Leverandør: Roth Carl
Gå til:
Product Details & Documents
Synonymer: 3-Hydroxy-4-(2-sulpho-4-[4-sulphophenylazo]phenylazo)-2,7-naphthalenedisulphonic acid sodium salt, CI 27195, Ponceau, 3-Hydroxy-4-((2-sulpho-4-((4-sulphophenyl)diazenyl)phenyl)diazenyl)naphthalene-2,7-disulphonic acid, tetrasodium salt
- CAS nummer:6226-79-5
- Formel:C₂₂H₁₂N₄Na₄O₁₃S₄
- MDL nummer:MFCD00003892
- Molecular Weight:760.56 g/mol
- Opbevaringstemperatur:Stue temperatur
- EINECS:228-319-2
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:10 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



