Ponceau S
Glass bottle for solids, 25 g
Katalog # ROTH5938.1
Marketsource
Leverandør: Roth Carl
Gå til:
Product Details & Documents
Synonymer: 3-Hydroxy-4-(2-sulpho-4-[4-sulphophenylazo]phenylazo)-2,7-naphthalenedisulphonic acid sodium salt, CI 27195, Ponceau, 3-Hydroxy-4-((2-sulpho-4-((4-sulphophenyl)diazenyl)phenyl)diazenyl)naphthalene-2,7-disulphonic acid, tetrasodium salt
- CAS nummer:6226-79-5
- Formel:C₂₂H₁₂N₄Na₄O₁₃S₄
- MDL nummer:MFCD00003892
- Molecular Weight:760.56 g/mol
- Opbevaringstemperatur:Stue temperatur
- EINECS:228-319-2
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:25 g



