Orders LinkContactUsLinkComponent
Borsyre
5 kg
  ROTH5935.2
Marketsource
 :   Roth Carl
Borsyre
  ROTH5935.2
 :  Roth Carl
Marketsource
 :  10043-35-3
Borsyre
Borsyre
5 kg
 :  10043-35-3
1 * 5 kg
 :  Boron trihydroxide, Boron hydroxide, Boric acid, Boracic acid
  •  :10043-35-3
  •  :B(OH)₃
  •  :MFCD00011337
  •  :61.83 g/mol
  •  :13,01326
  •  :185 °C (1013 hPa)
  •  :160 °C
  •  :1,44 g/cm³ (25 °C)
  •  :Stue temperatur
  •  :233-139-2
  •  :01-2119486683-25
  • Pakn:
    5 kg