Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Calciumhypochlorit

Calciumhypochlorit

Glass bottle for solids, 500 g
Katalog # ROTH5164.1
Oxiderende
Ætsende
Irriterende
Miljø
Marketsource
Leverandør:   Roth Carl
Calciumhypochlorit
Katalog # ROTH5164.1
Leverandør:  Roth Carl
Marketsource
CAS nummer:  7778-54-3
Calciumhypochlorit

Glass bottle for solids, 500 g

CAS nummer:  7778-54-3
Price
Spørg for pris
Log ind for tilgængelighed
1 * 500 g
Product Details & Documents
  • CAS nummer:7778-54-3
  • Formel:CaCl₂O₂
  • MDL nummer:MFCD00010900
  • Molecular Weight:142.98 g/mol
  • Merck Index:13,02111
  • Smeltende Pt:∼100 °C
  • Tæthed:2,35 g/cm³ (20 °C)
  • EINECS:231-908-7
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    500 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.