Methyl rød natrium salt
Glass bottle for solids, 25 g
ROTH5003.1
Marketsource
: Methyl red water soluble, 2-(4-Dimethylaminophenylazo)benzoic acid sodium salt, Sodium 2-(p-(dimethylamino)phenylazo)benzoate
- :845-10-3
- :C₁₅H₁₄N₃NaO₂
- :MFCD00002426
- :291.28 g/mol
- :13,06142
- :> 180 °C
- :Stue temperatur
- :212-682-9
- Pakning:Glass bottle for solids
- Pakn:25 g