Orders LinkContactUsLinkComponent
Methyl rød natrium salt
Methyl rød natrium salt
Glass bottle for solids, 25 g
  ROTH5003.1
Marketsource
 :   Roth Carl
Methyl rød natrium salt
  ROTH5003.1
 :  Roth Carl
Marketsource
 :  845-10-3
Methyl rød natrium salt
Glass bottle for solids, 25 g
 :  845-10-3
1 * 25 g
 :  Methyl red water soluble, 2-(4-Dimethylaminophenylazo)benzoic acid sodium salt, Sodium 2-(p-(dimethylamino)phenylazo)benzoate
  •  :845-10-3
  •  :C₁₅H₁₄N₃NaO₂
  •  :MFCD00002426
  •  :291.28 g/mol
  •  :13,06142
  •  :> 180 °C
  •  :Stue temperatur
  •  :212-682-9
  • Pakning:
    Glass bottle for solids
  • Pakn:
    25 g