Geraniol
Glass bottle, 100 mL
Catalog # ROTH4988.2
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: (E)-3,7-Dimethylocta-2,6-dien-1-ol
- CAS Number:106-24-1
- Formula:C₁₀H₁₈O
- MDL Number:MFCD00002917
- Molecular Weight:154.25 g/mol
- EINECS:203-377-1
Specifications
Specifications
- Pakning:Glass bottle
- Pakn:100 mL



