Geraniol
Glass bottle, 25 mL
Katalog # ROTH4988.1
Marketsource
Leverandør: Roth Carl
Gå til:
Product Details & Documents
Synonymer: (E)-3,7-Dimethylocta-2,6-dien-1-ol
- CAS nummer:106-24-1
- Formel:C₁₀H₁₈O
- MDL nummer:MFCD00002917
- Molecular Weight:154.25 g/mol
- EINECS:203-377-1
Specifikationer
Specifikationer
- Pakning:Glass bottle
- Pakn:25 mL



