Tetrachlorethylen
Glass bottle, 2,5 L
Katalog # ROTH4737.3
Marketsource
Leverandør: Roth Carl
Gå til:
Product Details & Documents
Synonymer: Tetrachloroethene, Ethylene tetrachloride, PCE, Perchloroethylene
- CAS nummer:127-18-4
- Formel:C₂Cl₄
- MDL nummer:MFCD00000834
- Molecular Weight:165.83 g/mol
- Merck Index:12,09332
- Kogende Pt:121,1 °C (1013 hPa)
- Smeltende Pt:–22 °C
- Tæthed:1,63 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:204-825-9
- FN:1897
- ADR:6.1,III
Specifikationer
Specifikationer
- Pakning:Glass bottle
- Pakn:2,5 L



