Tetrachlorethylen
Glass bottle, 1 L
ROTH4737.1
Marketsource
: Tetrachloroethene, Ethylene tetrachloride, PCE, Perchloroethylene
- :127-18-4
- :C₂Cl₄
- :MFCD00000834
- :165.83 g/mol
- :12,09332
- :121,1 °C (1013 hPa)
- :–22 °C
- :1,63 g/cm³ (20 °C)
- :Stue temperatur
- :204-825-9
- :1897
- :6.1,III
- Pakning:Glass bottle
- Pakn:1 L