Orders LinkContactUsLinkComponent
Tetrachlorethylen
Glass bottle, 1 L
  ROTH4737.1
Marketsource
 :   Roth Carl
Tetrachlorethylen
  ROTH4737.1
 :  Roth Carl
Marketsource
 :  127-18-4
Tetrachlorethylen
Tetrachlorethylen
Glass bottle, 1 L
 :  127-18-4
1 * 1 L
 :  Tetrachloroethene, Ethylene tetrachloride, PCE, Perchloroethylene
  •  :127-18-4
  •  :C₂Cl₄
  •  :MFCD00000834
  •  :165.83 g/mol
  •  :12,09332
  •  :121,1 °C (1013 hPa)
  •  :–22 °C
  •  :1,63 g/cm³ (20 °C)
  •  :Stue temperatur
  •  :204-825-9
  •  :1897
  •  :6.1,III
  • Pakning:
    Glass bottle
  • Pakn:
    1 L