Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

D(+)-Sucrose

5 kg
Katalog # ROTH4661.2
Marketsource
Leverandør:   Roth Carl
D(+)-Sucrose
Katalog # ROTH4661.2
Leverandør:  Roth Carl
Marketsource
CAS nummer:  57-50-1
D(+)-Sucrose
D(+)-Sucrose

5 kg

CAS nummer:  57-50-1
Price
Spørg for pris
Log ind for tilgængelighed
1 * 5 kg
Product Details & Documents
Synonymer:  β-ᴅ-Fructofuranosyl-α-ᴅ-glucopyranoside, α-ᴅ-Glucopyranosyl β-ᴅ-fructofuranoside, D(+)-Saccharose, α-ᴅ-Glc-(1→2)-β-ᴅ-Fru, Sucrose
  • CAS nummer:57-50-1
  • Formel:C₁₂H₂₂O₁₁
  • MDL nummer:MFCD00006626
  • Molecular Weight:342.3 g/mol
  • Merck Index:13,08966
  • Smeltende Pt:169…170 °C
  • Tæthed:1,29 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-334-9
Specifikationer

Specifikationer

  • Pakn:
    5 kg

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.