Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Transfer membranes, nitrocellulose

Nitrocellulose membranes, discs, pore size 0,45 µm, Ø 25 mm
Katalog # ROTH4589.1
Marketsource
Leverandør:   Roth Carl
Transfer membranes, nitrocellulose
Katalog # ROTH4589.1
Leverandør:  Roth Carl
Marketsource
Transfer membranes, nitrocellulose

Nitrocellulose membranes, discs, pore size 0,45 µm, Ø 25 mm

Price
Spørg for pris
Log ind for tilgængelighed
1 * 100 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Nitrocellulose membranes, discs, pore size 0,45 µm
  • Dimensioner:
    Ø 25 mm
  • Seq:
    05