Orders LinkContactUsLinkComponent
Magnetic stirring bars, triangular, PTFE-coated, Rotilabo®
Magnetic stirring bars, triangular, PTFE-coated, Rotilabo®
35 mm, 9 mm
Katalog # ROTH4557.1
Marketsource
Leverandør:   Roth Carl
Magnetic stirring bars, triangular, PTFE-coated, Rotilabo®
Katalog # ROTH4557.1
Leverandør:  Roth Carl
Marketsource
Magnetic stirring bars, triangular, PTFE-coated, Rotilabo®
35 mm, 9 mm
Price
Spørg for pris
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Ø:
    9 mm
  • Længde:
    35 mm