Eddikesyreanhydrid
Glass bottle, 1 L
Katalog # ROTH4483.1
Marketsource
Leverandør: Roth Carl
Gå til:
Product Details & Documents
Synonymer: Acetyl oxide, Acetyl acetate
- CAS nummer:108-24-7
- Formel:(CH₃CO)₂O
- MDL nummer:MFCD00008705
- Molecular Weight:102.09 g/mol
- Merck Index:13,00057
- Kogende Pt:139,9 °C (1013 hPa)
- Smeltende Pt:–73 °C
- Tæthed:1,08 g/cm³ (20 °C)
- Flash Pt:49 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:203-564-8
- FN:1715
- ADR:8,II
- NÅ:01-2119486470-36
Specifikationer
Specifikationer
- Pakning:Glass bottle
- Pakn:1 L
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



