Ninhydrin
Glass bottle for solids, 100 g
ROTH4378.2
Marketsource
: Ninhydrin monohydrate, Trioxohydrindene monohydrate, 2,2-Dihydroxyindane-1,3-dione, 2,2-Dihydroxy-1,3-indanedione, Indanetrione hydrate, 2,2-Dihydroxyindan-1,3-dion, 1,2,3-Indantrione monohydrate
- :485-47-2
- :C₉H₆O₄
- :MFCD00003791
- :178.14 g/mol
- :13,06582
- :351 °C (1013 hPa)
- :250…258 °C
- :0,862 g/cm³ (20 °C)
- :Stue temperatur
- :207-618-1
- Pakning:Glass bottle for solids
- Pakn:100 g