Tannin
Glass bottle for solids, 1 kg
Catalog # ROTH4239.2
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Gallotannin, Tannin
- CAS Number:1401-55-4
- Formula:C₇₆H₅₂O₄₆
- MDL Number:MFCD00066397
- Molecular Weight:1701.2 g/mol
- Merck Index:12,09221
- Melting Pt:218…219 °C
- Density:>1 g/cm³ (20 °C)
- Flash Pt:199 °C
- EINECS:215-753-2
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pakning:Glass bottle for solids
- Pakn:1 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



