Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
L(+)-Isoleucin

L(+)-Isoleucin

Glass bottle for solids, 100 g
Katalog # ROTH3922.2
Marketsource
Leverandør:   Roth Carl
L(+)-Isoleucin
Katalog # ROTH3922.2
Leverandør:  Roth Carl
Marketsource
CAS nummer:  73-32-5
L(+)-Isoleucin

Glass bottle for solids, 100 g

CAS nummer:  73-32-5
Price
Spørg for pris
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents
Synonymer:  L-Isoleucine, (2S,3S)-(+)-Isoleucine, H-Ile-OH
  • CAS nummer:73-32-5
  • Formel:H₃CCH₂CH(CH₃)CH(NH₂)COOH
  • MDL nummer:MFCD00064222
  • Molecular Weight:131.17 g/mol
  • Merck Index:14,05179
  • Smeltende Pt:279…280 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-798-2
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    100 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.