L(+)-Isoleucin
Glass bottle for solids, 10 g
Katalog # ROTH3922.1
Marketsource
Leverandør: Roth Carl
Gå til:
Product Details & Documents
Synonymer: L-Isoleucine, (2S,3S)-(+)-Isoleucine, H-Ile-OH
- CAS nummer:73-32-5
- Formel:H₃CCH₂CH(CH₃)CH(NH₂)COOH
- MDL nummer:MFCD00064222
- Molecular Weight:131.17 g/mol
- Merck Index:14,05179
- Smeltende Pt:279…280 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-798-2
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:10 g



