Chloramphenicol
Glass bottle for solids, 100 g
Catalog # ROTH3886.3
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Chloromycetin®, Tega-Cetin, (R,R)-Chloramphenicol
- CAS Number:56-75-7
- Formula:Cl₂CHCONHCH(CH₂OH)CH(OH)C₆H₄NO₂
- MDL Number:MFCD00078159
- Molecular Weight:323.13 g/mol
- Merck Index:13,02087
- Boiling Pt:645 °C (1013 hPa)
- Melting Pt:150 °C
- Density:1,49 g/cm³ (20 °C)
- Storage Temperature:Stue temperatur
- EINECS:200-287-4
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pakning:Glass bottle for solids
- Pakn:100 g



