Orders LinkContactUsLinkComponent
Chloramphenicol
Glass bottle for solids, 25 g
  ROTH3886.2
Marketsource
 :   Roth Carl
Chloramphenicol
  ROTH3886.2
 :  Roth Carl
Marketsource
 :  56-75-7
Chloramphenicol
Glass bottle for solids, 25 g
 :  56-75-7
1 * 25 g
 :  Chloromycetin®, Tega-Cetin, (R,R)-Chloramphenicol
  •  :56-75-7
  •  :Cl₂CHCONHCH(CH₂OH)CH(OH)C₆H₄NO₂
  •  :MFCD00078159
  •  :323.13 g/mol
  •  :13,02087
  •  :645 °C (1013 hPa)
  •  :150 °C
  •  :1,49 g/cm³ (20 °C)
  •  :Stue temperatur
  •  :200-287-4
  • Pakning:
    Glass bottle for solids
  • Pakn:
    25 g