Hæmatoxylin
Glass bottle for solids, 10 g
Leverandør: Roth Carl
Gå til:
Product Details & Documents
Synonymer: DcH-48, Hydroxybrazilin, (6aS,11bR)-6,6a,7,11b-Tetrahydroindeno[2,1-c]chromene-3,4,6a,9,10-pentaol, NSC 2785, Natural Black I, Hydroxybrasilin, (+)-Hematoxylin
- CAS nummer:517-28-2
- Formel:C₁₆H₁₄O₆
- MDL nummer:MFCD00078111
- Molecular Weight:302.28 g/mol
- Merck Index:13,04654
- Smeltende Pt:291…293 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:208-237-3
Certifikater
Søg efter partinummerSpecifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:10 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



