Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

L (+)-Glutaminsyre

Glass bottle for solids, 1 kg
Katalog # ROTH3774.2
Marketsource
Leverandør:   Roth Carl
L (+)-Glutaminsyre
Katalog # ROTH3774.2
Leverandør:  Roth Carl
Marketsource
CAS nummer:  56-86-0
L (+)-Glutaminsyre
L (+)-Glutaminsyre

Glass bottle for solids, 1 kg

CAS nummer:  56-86-0
Price
Spørg for pris
Log ind for tilgængelighed
1 * 1 kg
Product Details & Documents
Synonymer:  (S)-(+)-2-Aminopentanedioic acid, L(+)-1-Aminopropane-1,3-dicarboxylic acid, (S)-(+)-α-Glutamic acid, H-Glu-OH, L-Glutamic acid, L(+)-2-Aminopentanedioic acid, (S)-(+)-Glutamic acid, (S)-(+)-1-Aminopropane-1,3-dicarboxylic acid, L(+)-α-Glutamic acid
  • CAS nummer:56-86-0
  • Formel:HO₂CCH₂CH₂CH(NH₂)CO₂H
  • MDL nummer:MFCD00002634
  • Molecular Weight:147.13 g/mol
  • Merck Index:13,04482
  • Kogende Pt:325 °C (1013 hPa)
  • Smeltende Pt:160 °C
  • Tæthed:1,538 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-293-7
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    1 kg

Specifikationer