Lithiumchlorid
Glass bottle for solids, 1 kg
Katalog # ROTH3739.2
Marketsource
Leverandør: Roth Carl
Gå til:
Product Details & Documents
- CAS nummer:7447-41-8
- Formel:LiCl
- MDL nummer:MFCD00011078
- Molecular Weight:42.39 g/mol
- Merck Index:13,05550
- Kogende Pt:1382 °C (1013 hPa)
- Smeltende Pt:614 °C
- Tæthed:2,07 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:231-212-3
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:1 kg



