Eddikesyre
Glass bottle, 1 L
Katalog # ROTH3738.1
Marketsource
Leverandør: Roth Carl
Gå til:
Product Details & Documents
Synonymer: Vinegar acid, Methanecarboxylic acid, Ethanoic acid
- CAS nummer:64-19-7
- Formel:H₃CCOOH
- MDL nummer:MFCD00036152
- Molecular Weight:60.05 g/mol
- Kogende Pt:118 °C (1013 hPa)
- Smeltende Pt:17 °C
- Tæthed:1,05 g/cm³ (20 °C)
- Flash Pt:38,5 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-580-7
- FN:2789
- ADR:8,II
- NÅ:01-2119475328-30
Specifikationer
Specifikationer
- Pakning:Glass bottle
- Pakn:1 L